Cole-Parmer® RH-400 Series Integrated Heated Circulating Baths with MX Temperature Controller
Simple to install, set up and use, with a swiveling controller and easy-to-see digital display
  • Backlit controller display shows actual temperature and set point
  • Temperatures stay constant due to intelligent PID algorithms and a variable-speed pump
  • Three tank sizes from 7 to 20 L
  • Operates at low noise levels
Designed for almost any laboratory or process application, the heated baths are easy to use and offer precise temperature control. The top plate stays cool at high temperatures and can be easily cleaned and disinfected. Rear inlet/outlet ports enable use in external process applications. Standard housing on the 7 L, 15 L and 20 L models have chemical-resistant stainless steel tanks with reservoir covers and a drain and port behind the front access panel. The polycarbonate open bath systems (17 L, 28 L) have a cooling coil opening, reservoir drain port and offer excellent visibility thanks to the clear housing.
£2,100.01 - £2,652.47GBP / Each
£2,520.01 - £3,182.96GBP (incl VAT)
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (L/min) Power (VAC) Reservoir Capacity (L)
Availability Pricing
COMPARE
13.5 120 7
£2,100.01GBP / Each
£2,520.01GBP (incl VAT)
COMPARE
11.9 240 7
£2,100.01GBP / Each
£2,520.01GBP (incl VAT)
COMPARE
13.5 120 15
£2,548.62GBP / Each
£3,058.34GBP (incl VAT)
COMPARE
13.5 120 20
£2,652.47GBP / Each
£3,182.96GBP (incl VAT)