This product is now discontinued, View Related Products
Cole-Parmer® Polystat® Low-Temperature Benchtop Chillers; -20° to +40°C
Cool and compact, with the power you need
  • Enhance productivity with minimal noise output and a small footprint
  • Capable of cooling two rotary evaporators concurrently
  • Large, easy-to-read LED
  • Illuminated fluid-level indicator for easy viewing
Get the job done, conveniently, quietly and efficiently. These sleek recirculating chillers feature a top-mounted fill port with built-in fluid filter and 0.7 gal. (2.65 L) reservoir. Set temperatures from –20 to 40°C on the easy-to-use control panel. PID temperature controller offers stability of ±0.1°C. Adjustable high-temperature limit, alarm restart on power failure and a low flow rate alarm with power cutoff provide important safeguards.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Cooling Capacity At 0° C (watts) Max Flow Rate (L/min) Power (VAC)
Availability Pricing
COMPARE
680 14.8 120
Call For Price
COMPARE
680 12.9 240