Cole-Parmer® PB-350 Series High-Capacity Portable Toploading Balances
Easily weigh heavy dynamic loads
  • Included software collects and exports data for further analysis
  • Built-in rechargeable battery provides portability
  • Connect to printer or PC with RS-232 interface
  • Weighing modes include parts counting, percent weighing, totalizing, check weighing, peak hold, and animal weighing
  • 5-year industry-leading warranty
These toploading balances feature high weighing capacities and the ability to weigh moving samples, such as live animals. Included data collection software allows you to export logged data for further analysis, and RS-232 interface provides printer or PC connection. Balances may run on mains power, or use the built-in rechargeable battery where portability is necessary.
£852.40 - £1,358.48GBP / Each
£1,022.88 - £1,630.18GBP (incl VAT)
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Power (VAC)
Availability Pricing
COMPARE
External 1000 100 to 240
£852.40GBP / Each
£1,022.88GBP (incl VAT)
COMPARE
External 2000 100 to 240
£972.60GBP / Each
£1,167.12GBP (incl VAT)
COMPARE
External 6000 100 to 240
£881.41GBP / Each
£1,057.69GBP (incl VAT)
COMPARE
External 10000 100 to 240
£1,071.97GBP / Each
£1,286.36GBP (incl VAT)
COMPARE
External 20000 100 to 240
£1,358.48GBP / Each
£1,630.18GBP (incl VAT)
COMPARE
Internal 1000 100 to 240
£1,053.23GBP / Each
£1,263.88GBP (incl VAT)
COMPARE
Internal 2000 100 to 240
£1,098.75GBP / Each
£1,318.50GBP (incl VAT)
COMPARE
Internal 6000 100 to 240
£1,189.60GBP / Each
£1,427.52GBP (incl VAT)
Showing 1- 8 of 8