Cole-Parmer Panel-Mount Solar Powered LED Thermometers
Perfect for use in freezers and walk-in coolers!
  • Replace existing 2" mechanical thermometers!
  • NSF (National Sanitation Foundation) certified for food storage areas
These maintenance-free thermometers operate on solar power in light levels above 100 LUX, and use a battery back-up below 100 LUX. The calibration adjustment is on back of unit. The 48"L cable has no lead-containing pigments.
Use panel-mount thermometers for front flange attachment.
Use wall-mount thermometers for rear flange attachment.
U-clamp thermometers are for mounting without flange attachment.
£46.00 - £79.58GBP / Each
£55.20 - £95.50GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Display Type Max Temperature (° F) Min Temperature (° F)
Availability Pricing
COMPARE
158 -40
£46.00GBP / Each
£55.20GBP (incl VAT)
COMPARE
3.5-digit LCD 158 -40
£79.58GBP / Each
£95.50GBP (incl VAT)