Cole-Parmer® INC-200 Series Mini Digital Incubators
Eliminate the need for external thermometers
  • Large internal chamber accepts bottles and flasks up to 2 L
  • Built-in internal outlet powers small shakers and rockers
  • See-through panel lets you view entire chamber without opening door
These compact incubators with digital temperature control eliminate the need for external thermometers and repetitive “fine tuning” of an analog controller. The incubators immediately begin to heat up or cool down (depending on the model) as soon as the desired temperature is set. The chamber temperature is continuously monitored, and the real-time temperature can be easily viewed on the large LED display
£566.22 - £874.72GBP / Each
£679.46 - £1,049.66GBP (incl VAT)
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (cu ft) Power (VAC) Description
Availability Pricing
COMPARE
0.7 115 Mini Digital Incubator, Heat Only, 115 V
£566.22GBP / Each
£679.46GBP (incl VAT)
COMPARE
0.7 115 Mini Digital Incubator, Heat and Cool, 115 V
£874.72GBP / Each
£1,049.66GBP (incl VAT)
COMPARE
0.7 230 Mini Digital Incubator, Heat and Cool, 230 V
£874.72GBP / Each
£1,049.66GBP (incl VAT)
COMPARE
0.7 230 Mini Digital Incubator, Heat Only, 230 V
£566.22GBP / Each
£679.46GBP (incl VAT)