Cole-Parmer® HP-200 Series Large-Capacity Infrared Hot Plates
Efficient infrared heater saves you time and energy
  • Large plate surface is ideal for heating large volumes of liquid
  • Ceramic top stays cool around the edges, preventing burns
  • Be safe—flashing light warns when top plate is hot
  • BioCote® antimicrobial protection is built in for a cleaner, more hygienic product
This hot plate features an efficient, 900-watt infrared heater that can boil a liter of water over 30% faster than a conventional, 1200-watt ceramic hot plate. "Hot" warning light flashes when the plate temperature is above 50 °C and remains lit even when the hot plate is turned off.
Units feature BioCote antimicrobial protection which reduces the presence of odor and stain causing microbes such as bacteria, mold, and fungi on the product surface by up to 99.99%.
£914.12GBP / Each
£1,096.94GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Power (VAC) Top Plate Length (in)
Availability Pricing
COMPARE
842 230 12
£914.12GBP / Each
£1,096.94GBP (incl VAT)
COMPARE
842 120 12
£914.12GBP / Each
£1,096.94GBP (incl VAT)