This product is now discontinued, View Related Products
Cole-Parmer Horizontal Mid-Size Gel Trays
For Gel system 28560-00
Gel Trays let you pour gels in the unit using the tapered baffles; outside of the unit by taping the ends of the tray; or outside of the unit using the gel casters sold seperately. Trays are UV-transparent acrylic. Multiple comb slots let you cast gels for simultaneous short runs.
With 4 comb slots; pathlength of 3.1 cm/slot
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Gel Tray for Horizontal Mid-Size Gel System Model 28560-00
Call For Price
COMPARE
Gel Tray for Horizontal Mid-Size Gel System Model 28560-10