Cole-Parmer High-Form Crucibles & Covers, Porcelain
High-form crucibles are glazed inside and out except for outside bottom. Withstands temperatures to 2102°F (1150°C). Cover is included.
£11.20 - £16.20GBP / Each
£13.44 - £19.44GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL)
Availability Pricing
COMPARE
15
£11.20GBP / Each
£13.44GBP (incl VAT)
COMPARE
25
£14.06GBP / Each
£16.87GBP (incl VAT)
COMPARE
50
£16.20GBP / Each
£19.44GBP (incl VAT)