This product is now discontinued, View Related Products
Foxx Life Sciences High-Flow Bottletop Filtration Systems
Eliminate leakage with molded one-piece funnel design
  • Extra space in the bottle allows up to a 15% dilution
  • Polystyrene funnel and bottle have large raised graduations for easy reading
  • Entire assembly is gamma sterilized
  • Stable, hands-free filtering with optional pedestal or cradle ring
Large-diameter 0.2 µm PES membrane covers the entire round bottom of the funnel, allowing you to obtain high throughput and fast flow rates. Cap features write-on surface and gripping area for easy handling. Receiving bottle also features molded-in finger grips.
Connect the funnel to a vacuum hose in three separate ways. Use the included vacuum hose adapter to make a direct connection. Order the optional pedestal or cradle ring to create a convenient docking station for hands-free filtering.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Pore Size (µm) Description
Availability Pricing
COMPARE
1000 0.2 High Speed Bottle Top Filtration Unit, Full Assembly, 1000 mL, 0.2um
Call For Price
Foxx Life Sciences High Speed Bottle Top Filtration Unit, Full Assembly, 0.2 um; 250 mL WZ-06060-57 Mfr # 116-7233-OEM
COMPARE
250 0.2 High Speed Bottle Top Filtration Unit, Full Assembly, 250 mL, 0.2um
Call For Price
Foxx Life Sciences High Speed Bottle Top Filtration Unit, Full Assembly, 0.2 um; 500 mL WZ-06060-58 Mfr # 116-8233-RLS
COMPARE
500 0.2 High Speed Bottle Top Filtration Unit, Full Assembly, 500 mL, 0.2um
Call For Price
COMPARE
1000 0.2 High Speed Bottle Top Filtration Unit, Funnel Only, 1000 mL, 0.2um
Call For Price
COMPARE
250 0.2 High Speed Bottle Top Filtration Unit, Funnel Only, 250 mL, 0.2um
Call For Price
COMPARE
500 0.2 High Speed Bottle Top Filtration Unit, Funnel Only, 500 mL, 0.2 um