This product is now discontinued, View Related Products
Cole-Parmer® Essentials Heavy-Duty Laboratory Frame Sets
Corrosion resistant frames and rods
  • Custom fit any laboratory with the right frame kit for your application
Heavy-Duty Lab-Frame
Vertical/floor mounting, Free standing unit is designed specifically for heavy-duty work such as pilot plant set-ups. The entire frame is supported on sturdy, counter-weighted cast iron feet, which may be bolted to the floor for added stability.
Frame only dimensions (W x H): 48 x 72 (121.9 x 182.9 cm)
Overall Dimensions (W x H): 75 x 72” (190.5 x 182.9 cm)
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Heavy Duty Lab Frame, Aluminum, 72\" W x 75\" H x 21.75\" D
Call For Price
COMPARE
Heavy-Duty Lab Frame, Stainless Steel, 72\" W x 75\" H x 21-3/4\" D
Call For Price
COMPARE
Heavy-Duty Lab Frame, Fiberglass, 72\" W x 75\" H x 21-3/4\" D