Cole-Parmer® Essentials Heavy-Duty Carboys with Shoulder Handle and Spigots, Polypropylene
Sturdy, octagonal design for added rigidity
  • Easier filling and pouring with offset neck
  • Leakproof spigot models have tube fitting for easier filling of containers
  • Polypropylene closure reduces evaporation
  • Autoclavable
Heavy-duty graduated polypropylene (PP) carboys feature octagonal shape for increased rigidity when full. The neck is offset for easier filling and pouring. Molded handle is sealed at the ends to prevent contamination. Durable, leakproof spigot is supplied with a tube fitting to accept 0.31" (8-mm) ID tubing. Carboys include a PP screw closure. Autoclavable up to 275°F.
£112.60 - £146.55GBP / Pkg of 1
£135.12 - £175.86GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Capacity (L) Height (in) Material
Availability Pricing
COMPARE
5 10 5/8 Polypropylene(PP)
£112.60GBP / Pkg of 1
£135.12GBP (incl VAT)
COMPARE
10 14 1/8 Polypropylene(PP)
£124.85GBP / Pkg of 1
£149.82GBP (incl VAT)
COMPARE
20 16 1/2 Polypropylene(PP)
£146.55GBP / Pkg of 1
£175.86GBP (incl VAT)