Generator Probes for Cole-Parmer® HO-200 and HO-400 Series Homogenizers
Choose the generator based on the dispersion, tissue type, and or sample size
  • Applications include large volume emulsions, suspensions, and dispersions for liquid/liquid or solid/liquid
Select from stainless steel (SS) or disposable polycarbonate (PC) generators. Stainless steel probes are ideal for larger samples. Disposable probes are ideal for when cross contamination may be of concern.
£208.86 - £2,510.31GBP / Each
£250.63 - £3,012.37GBP (incl VAT)
17 variations of this product are available.
See All Product Options Below
17 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Max Sample Size (mL) Min Sample Size (mL)
Availability Pricing
COMPARE
Stainless steel 30 0.25
£1,453.29GBP / Each
£1,743.95GBP (incl VAT)
COMPARE
Stainless steel 30 0.25
£1,398.59GBP / Each
£1,678.31GBP (incl VAT)
COMPARE
Stainless steel 100 1.5
£1,600.21GBP / Each
£1,920.25GBP (incl VAT)
COMPARE
Stainless steel 100 1.5
£1,507.55GBP / Each
£1,809.06GBP (incl VAT)
COMPARE
Stainless steel 2000 100
£1,892.06GBP / Each
£2,270.47GBP (incl VAT)
COMPARE
Stainless steel 2000 100
£2,036.99GBP / Each
£2,444.39GBP (incl VAT)
COMPARE
Stainless steel 5000 200
£2,475.76GBP / Each
£2,970.91GBP (incl VAT)
COMPARE
Stainless steel 5000 200
£2,510.31GBP / Each
£3,012.37GBP (incl VAT)
COMPARE
Stainless steel 5 0.05
£1,369.34GBP / Each
£1,643.21GBP (incl VAT)
COMPARE
Stainless steel 5 0.05
£1,335.60GBP / Each
£1,602.72GBP (incl VAT)
COMPARE
Polycarbonate 30 0.25
£414.16GBP / Pkg of 25
£496.99GBP (incl VAT)
COMPARE
Polycarbonate 30 0.25
£766.36GBP / Pkg of 50
£919.63GBP (incl VAT)
COMPARE
Polycarbonate 30 0.25
£406.47GBP / Pkg of 25
£487.76GBP (incl VAT)
COMPARE
Polycarbonate 30 0.25
£709.09GBP / Pkg of 50
£850.91GBP (incl VAT)
COMPARE
Stainless steel, Ultem 30 0.25
£612.83GBP / Each
£735.40GBP (incl VAT)
COMPARE
Ultem 30 0.25
£208.86GBP / Each
£250.63GBP (incl VAT)
COMPARE
Polycarbonate 30 0.25
£408.48GBP / Pkg of 25
£490.18GBP (incl VAT)
Showing 1- 17 of 17