This product is now discontinued, View Related Products
Cole-Parmer Desiccant-Filled Plates
  • Silica gel-filled poylystyrene plates
Replace bulk desiccant with these silica gel-filled plates. The clear polystyrene plates let you visually inspect for color changes from blue to pink as it absorbs moisture. Regenerate desiccant by heating plates in a 122°F (50°C) oven overnight. Plates are individually sealed.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Plate Size (mm)
Availability Pricing
Bel-Art 42048-8100 Desiccant-Filled Plates; 100-mm diameter; 6/pack WZ-07193-90 Mfr # 42048-8100
COMPARE
100
Call For Price
Bel-Art Desiccant-Filled Plates; 65-mm diameter; 6/PK WZ-07193-95 Mfr # 42048-8065
COMPARE
65