This product is now discontinued, View Related Products
Cole-Parmer® Carboy Safety Funnels, SS
Improve the safety of your solvent waste system with this safety funnel
  • Constructed of high-quality, chemical-resistant stainless steel for enhanced durability
  • Overfill protection stops spillage of liquids during filling
  • Attached lid keeps volatile organic compounds (VOCs) from evaporating into the air
  • Removable, easy-to-clean flame trap prevents liquid waste from igniting
  • Available with 83B or 120 mm polypropylene carboy cap
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Closure Size (mm) Material
Availability Pricing
COMPARE
83 Stainless steel
Call For Price
COMPARE
120 Stainless steel