This product is now discontinued, View Related Products
Cole-Parmer® Ultrasonic Cleaners with Heater, Timer, and Power Boost
Protect sensitive items—safety shutoff and temperature-selectable alarm light for units with heat
  • Easily preselect run time and temperature with rotary dials
  • Obtain uniform sound waves and cleaning from the sweep function
  • Activate the pulse function for 20% increase in ultrasonic power and soil removal
The analog series of ultrasonic cleaners feature simple control with superior cleaning technology. Select heating temperature from 30 to 80°C, in 5° increments, with a rotary dial. Choose a maximum temperature for sensitive items and instruments—LED light illuminates if reached. Heating is not available on items 59989-29 and -30. The cleaners can be set to operate continuously or in timed runs, from 1 to 30 minutes (dial-selected). The integrated sweep mode ensures even sound distribution throughout the bath at 37 kHz for efficient and effective cleaning. Turn on pulse mode to increase ultrasonic power 20%—ideal for those hard to remove soils and pastes. Safety features include auto-shutoff if running for 8 hours continuously or if bath temperature exceeds 90°C, and a sealed display and elevated feet to protect against splashes and spills. Baths with capacities above 1 gallon also feature a side-mounted drain for easy emptying and cleaning.
£510.32GBP / Each
£612.38GBP (incl VAT)
20 variations of this product are available.
See All Product Options Below
20 PRODUCT OPTIONS
Compareitem specifications
Capacity (Gal) Power (VAC) Description
Availability Pricing
COMPARE
0.25 120 Ultrasonic Cleaner, 0.25 gal, 120 VAC
Call For Price
COMPARE
0.25 120 Ultrasonic Cleaner, 0.25 gal, 220 VAC
Call For Price
COMPARE
0.25 120 Ultrasonic Cleaner with Heat, 0.25 gal, 120 VAC
£510.32GBP / Each
£612.38GBP (incl VAT)
COMPARE
0.25 220 Ultrasonic Cleaner with Heat, 0.25 gal, 220 VAC
Call For Price
COMPARE
0.5 120 Ultrasonic Cleaner with Heat, 0.5 gal, 120 VAC
Call For Price
COMPARE
0.5 220 Ultrasonic Cleaner with Heat, 0.5 gal, 220 VAC
Call For Price
COMPARE
0.75 120 Ultrasonic Cleaner with Heat, 0.75 gal, 120 VAC
Call For Price
COMPARE
0.75 220 Ultrasonic Cleaner with Heat, 0.75 gal, 220 VAC
Call For Price
COMPARE
1.25 120 Ultrasonic Cleaner with Heat, 1.25 gal, 120 VAC
Call For Price
COMPARE
1.25 220 Ultrasonic Cleaner with Heat, 1.25 gal, 220 VAC
Call For Price
COMPARE
1.5 120 Ultrasonic Cleaner with Heat, 1.5 gal, 120 VAC
Call For Price
COMPARE
1.5 220 Ultrasonic Cleaner with Heat, 1.5 gal, 220 VAC
Call For Price
COMPARE
2.5 120 Ultrasonic Cleaner with Heat, 2.5 gal, 120 VAC
Call For Price
COMPARE
2.5 220 Ultrasonic Cleaner with Heat, 2.5 gal, 220 VAC
Call For Price
COMPARE
3.5 120 Ultrasonic Cleaner with Heat, 3.5 gal, 120 VAC
Call For Price
COMPARE
3.5 220 Ultrasonic Cleaner with Heat, 3.5 gal, 220 VAC
Call For Price
COMPARE
4.75 120 Ultrasonic Cleaner with Heat, 4.75 gal, 120 VAC
Call For Price
COMPARE
4.75 220 Ultrasonic Cleaner with Heat, 4.75 gal, 220 VAC
Call For Price
COMPARE
7.5 120 Ultrasonic Cleaner with Heat, 7.5 gal, 120 VAC
Call For Price
COMPARE
7.5 220 Ultrasonic Cleaner with Heat, 7.5 gal, 220 VAC
Call For Price
Showing 1- 20 of 20