This product is now discontinued, View Related Products
Cole-Parmer Alcohol Proof Glass Hydrometers
Measure percent of alcohol in various spirits
  • Temperature standard is 60°F (15.6°C)
  • Individually serialized for easy identification
  • Ensure accuracy with multi-language statement of accuracy supplied by manufacturer
  • All hydrometers are 100% lead free
Alcohol proof precision hydrometers measure the percent of alcohol in various spirits during distilling and rectifying. These hydrometers provide scales from 0 to 200% with 0.1% divisions.
Call For Price
20 variations of this product are available.
See All Product Options Below
20 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
0/11 Percent Alcohol Proof Precision Hydrometer
Call For Price
COMPARE
9/20 Percent Alcohol Proof Precision Hydrometer
Call For Price
COMPARE
19/30 Percent Alcohol Proof Precision Hydrometer
Call For Price
COMPARE
29/40 Percent Alcohol Proof Precision Hydrometer
Call For Price
COMPARE
39/50 Percent Alcohol Proof Precision Hydrometer
Call For Price
COMPARE
49/60 Percent Alcohol Proof Precision Hydrometer
Call For Price
COMPARE
59/70 Percent Alcohol Proof Precision Hydrometer
Call For Price
COMPARE
69/80 Percent Alcohol Proof Precision Hydrometer
Call For Price
COMPARE
79/90 Percent Alcohol Proof Precision Hydrometer
Call For Price
COMPARE
89/100 Percent Alcohol Proof Precision Hydrometer
Call For Price
COMPARE
99/110 Percent Alcohol Proof Precision Hydrometer
Call For Price
COMPARE
109/120 Percent Alcohol Proof Precision Hydrometer
Call For Price
H-B Instrument B61807-6300 DURAC 119/130 Percent Alcohol Proof Precision Hydrometer WZ-90310-43 Mfr # B61807-6300
COMPARE
119/130 Percent Alcohol Proof Precision Hydrometer
Call For Price
H-B Instrument B61807-6400 DURAC 129/140 Percent Alcohol Proof Precision Hydrometer WZ-90310-44 Mfr # B61807-6400
COMPARE
129/140 Percent Alcohol Proof Precision Hydrometer
Call For Price
COMPARE
139/150 Percent Alcohol Proof Precision Hydrometer
Call For Price
COMPARE
149/160 Percent Alcohol Proof Precision Hydrometer
Call For Price
COMPARE
159/170 Percent Alcohol Proof Precision Hydrometer
Call For Price
H-B Instrument B61807-6800 DURAC 169/180 Percent Alcohol Proof Precision Hydrometer WZ-90310-48 Mfr # B61807-6800
COMPARE
169/180 Percent Alcohol Proof Precision Hydrometer
Call For Price
COMPARE
179/190 Percent Alcohol Proof Precision Hydrometer
Call For Price
COMPARE
189/200 Percent Alcohol Proof Precision Hydrometer
Call For Price
Showing 1- 20 of 20