This product is now discontinued, View Related Products
Clamps & Accessories for Applikon RAMbio Benchtop Acoustic Mixer
Optimize performance of your Applikon RAMbio® Benchtop Acoustic Mixer
Mixer accommodates flasks ranging from 250 mL to 1 L (order mixer 29206-83 separately).
Use the RAMbio mixer in conjunction with the Oxy-Pump stopper to achieve bioreactor-equivalent oxygen transfer. The stopper actively pumps air into and out of your flask cultures. It offers complete headspace refreshment equivalent to that achieved in stirred tank bioreactors without the need for mechanical pumps. This autoclavable stopper is constructed from silicone and polypropylene and has a replaceable filter.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Applikon RAMbio Z861025010 Clamp for 250 mL Flask, with screws WZ-29206-84 Mfr # Z861025010
COMPARE
Clampfor250mLFlask,withscrews
Call For Price
Applikon RAMbio Z861050010 Clamp for 500 mL Flask, with screws WZ-29206-85 Mfr # Z861050010
COMPARE
Clampfor500mLFlask,withscrews
Call For Price
Applikon RAMbio Z861100010 Clamp for 1000 mL Flask, with screws WZ-29206-86 Mfr # Z861100010
COMPARE
Clampfor1000mLFlask,withscrews
Call For Price
Applikon RAMbio Z861700010 Strobe Assembly Kit for RAMbio WZ-29206-87 Mfr # Z861700010
COMPARE
Strobe Assembly Kit for RAMbio
Call For Price
Applikon RAMbio Z861800010 Pedestal Base, 18" with Drawer WZ-29206-88 Mfr # Z861800010
COMPARE
PedestalBase,18\"withDrawer
Call For Price
Applikon Z861900010 OxyPump Complete Stopper Assembly (pack of 10) WZ-29206-89 Mfr # Z861900010
COMPARE
OxyPump Complete Stopper Assembly (pack of 10)