This product is now discontinued, View Related Products
Chemware PTFE Policemen
PTFE Policeman has chisel- and diagonal-cut tips for scraping. Max temperature is 550°F (287°C). Autoclavable.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Material Sterile Description
Availability Pricing
Chemware D1069093 PTFE Policemen/Stir Rod, 6" L; 1/Pk WZ-06369-01 Mfr # D1069093
COMPARE
PTFE No PTFE Policemen/Stir Rod, 6\" L; 1/Pk
Call For Price
Chemware D1069094 PTFE Policemen/Stir Rod, 8" L; 1/Pk WZ-06369-02 Mfr # D1069094
COMPARE
PTFE No PTFE Policemen/Stir Rod, 8\" L; 1/Pk