Chemically Inert Two-Way Ball Valves
Safely control flow in chemically aggressive applications
  • Constructed of chemically inert PFA
All wetted materials are PFA—ideal for use with corrosive chemicals. Simply make a 1/4-turn to start or stop the flow. Maximum pressure is 150 psi; (10.3 bar) fluid temperature range is 70 to 400°F (21 to 204°C); operating temperature range is 0 to 150°F (–18 to 66°C).
£372.71 - £525.48GBP / Each
£447.25 - £630.58GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Inlet Port Orifice (in) Outlet Port
Availability Pricing
COMPARE
1/4" NPT(F) 0.25 1/4" NPT(F)
£372.71GBP / Each
£447.25GBP (incl VAT)
COMPARE
1/2" NPT (F) 0.375 1/2" NPT (F)
£525.48GBP / Each
£630.58GBP (incl VAT)