This product is now discontinued, View Related Products
Centrifan PE Small Volume Evaporators
Simply set temperature and timer, close the cover and walk away--no monitoring required
  • Ideal to concentrate small samples directly in vials
  • Small foot-print easily fits on benchtop or a lab cart
  • No vacuum pump--avoid bumping and potential sample cross-contamination
  • Green product --uses less energy, and designed for less exposure to toxic vapors
The Centrifan PE is ideal for concentrating small samples in research, pharmaceutical, and biotech labs working with DNA, RNA, and proteins. The proprietary self-generated blow-down technology protects samples from loss and it eliminates the potential problems with standard evaporators such as vacuum centrifuges, rotary evaporators, and freezer dryers. The unit has an adjustable temperature set point up to 55°C, cold trap, and a manual on/off control. Models also available with a digital timer.
The flexible Cetrifan PE has seven standard rotors to fit your applications. Rotors are available to accommodate vessels ranging from 1.5-ml HPLC vials up to 40-ml scintillation vials
.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
KD Scientific Centrifan PE-T, Small Volume Evaporator with cold trap and timer, 115V WZ-28690-02 Mfr # 78-0070
COMPARE
115
Call For Price
KD Scientific Centrifan PE, Small Volume Evaporator with cold trap, 115V WZ-28690-04 Mfr # 78-0071
COMPARE
115
Call For Price
KD Scientific Centrifan PE-T, Small Volume Evaporator with cold trap and timer, 230V WZ-28690-06 Mfr # 78-1070
COMPARE
230
Call For Price
KD Scientific Centrifan PE, Small Volume Evaporator with cold trap, 230V WZ-28690-08 Mfr # 78-1071
COMPARE
230