CAPP® ExpellPlus 384 Multichannel Pipettor Pipette Tips
Presterilized tips save time and reduce cross-contamination
  • Compatible with the CAPP® CAPPAero384 multichannel pipettor
Specially engineered to fit CappAero™ multichannel pipettors. ExpellPlus tips are suitable for CAPP 16, 24, 32, 48 and 64 channel pipettes. These high-accuracy, low-retention tips are designed to minimize sample loss due to stickier substances like DNA, RNA, and proteins.
£514.12 - £575.30GBP / Case of 10
£616.94 - £690.36GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Filter Type Max Volume (µL) Sterile
Availability Pricing
Capp ExpellPlus Pipette Tips, 0.2-50 µL, Sterile, No Filter; 384/Rack, 10 Racks/Cs WZ-25019-32 Mfr # 5030005C
COMPARE
None 50 Yes
£514.12GBP / Case of 10
£616.94GBP (incl VAT)
Capp ExpellPlus Pipette Tips, 0.2-50 µL, Sterile, Filtered; 384/Rack, 10 Racks/Pk WZ-25019-35 Mfr # 5030006
COMPARE
Polyethylene 50 Yes
£575.30GBP / Pkg of 10
£690.36GBP (incl VAT)