Cannon®-Fenske Calibrated Glass Capillary Routine Viscometers
Determine viscosities using ASTM testing methods
  • Each viscometer is calibrated in accordance with ASTM D446 Comes with a calibration certificate that includes the viscometer constant and formulas for calculating viscosities
Quickly and easily measure the viscosities of transparent Newtonian liquids in the chemical and petroleum industries. Require a 7-mL sample volume.
£178.74 - £183.65GBP / Each
£214.49 - £220.38GBP (incl VAT)
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
Compareitem specifications
Approximate Constant (cSt/sec) Max Viscosity Range (cSt) Universal Size Number
Availability Pricing
Cannon 9721-B50 Viscometer, universal size 25 WZ-98934-50 Mfr # 9721-B50
COMPARE
0.002 2 25
£183.65GBP / Each
£220.38GBP (incl VAT)
Cannon 9721-B53 Viscometer, universal size 50 WZ-98934-51 Mfr # 9721-B53
COMPARE
0.004 4 50
£183.65GBP / Each
£220.38GBP (incl VAT)
Cannon 9721-B56 Viscometer, universal size 75 WZ-98934-52 Mfr # 9721-B56
COMPARE
0.008 8 75
£183.65GBP / Each
£220.38GBP (incl VAT)
Cannon 9721-B59 Viscometer, universal size 100 WZ-98934-53 Mfr # 9721-B59
COMPARE
0.015 15 100
£183.65GBP / Each
£220.38GBP (incl VAT)
Cannon 9721-B62 Viscometer, universal size 150 WZ-98934-54 Mfr # 9721-B62
COMPARE
0.035 35 150
£183.65GBP / Each
£220.38GBP (incl VAT)
Cannon 9721-B65 Viscometer, universal size 200 WZ-98934-55 Mfr # 9721-B65
COMPARE
0.1 100 200
£183.65GBP / Each
£220.38GBP (incl VAT)
Cannon 9721-B68 Viscometer, universal size 300 WZ-98934-56 Mfr # 9721-B68
COMPARE
0.25 250 300
£183.65GBP / Each
£220.38GBP (incl VAT)
Cannon 9721-B71 Viscometer, universal size 350 WZ-98934-57 Mfr # 9721-B71
COMPARE
0.5 500 350
£183.65GBP / Each
£220.38GBP (incl VAT)
Cannon 9721-B74 Viscometer, universal size 400 WZ-98934-58 Mfr # 9721-B74
COMPARE
1.2 1200 400
£183.65GBP / Each
£220.38GBP (incl VAT)
Cannon 9721-B77 Viscometer, universal size 450 WZ-98934-59 Mfr # 9721-B77
COMPARE
2.5 2500 450
£183.65GBP / Each
£220.38GBP (incl VAT)
Cannon 9721-B80 Viscometer, universal size 500 WZ-98934-60 Mfr # 9721-B80
COMPARE
8 8000 500
£183.65GBP / Each
£220.38GBP (incl VAT)
Cannon 9721-B83 Viscometer, universal size 600 WZ-98934-61 Mfr # 9721-B83
COMPARE
20 20000 600
£183.65GBP / Each
£220.38GBP (incl VAT)
Cannon 9721-B86 Viscometer, universal size 650 WZ-98934-62 Mfr # 9721-B86
COMPARE
45 45000 650
£183.65GBP / Each
£220.38GBP (incl VAT)
Cannon 9721-B89 Viscometer, universal size 700 WZ-98934-63 Mfr # 9721-B89
COMPARE
100 100000 700
£178.74GBP / Each
£214.49GBP (incl VAT)
Showing 1- 14 of 14