This product is now discontinued, View Related Products
Cal Control 9500-Series Temperature/Process Controllers
Programmable profiling / autotuning P.I.D. industrial controllers.
  • IP66- and NEMA-rated 4X faceplate allows the controller to be used in washdown applications
  • Retrofittable communication RS232 or RS485
  • Available in 1/16-DIN size
These controllers can be configured in a wide range of temperature or process control options allowing the controller to be dedicated to a specific application — ideal for both OEM and manufacturing process applications. This combination of programmable ramp/soak profiles, process control inputs and outputs, along with optional communications makes the CAL 9500 a unique and affordable product. The programmable profiling capabilities of the controller allow for up to 31 programs (profiles) and up to 126 segments, unlimited use of event outputs, and up to 999 program loop cycles or continuous loop cycles. The Copy/Paste/Edit/Delete functions simplify program building. Other programmable features allow you to use sub-programs and to set three power-fail recovery options (Hold, Continue or Reset). The Hold back function ensures the next segment is not started until the last segment reaches the set point. These controllers also feature a single ramp/soak (dwell) program, five alarm modes, multilevel security lockout, and heat-cool operation. A mounting sleeve allows for easy mounting, maintenance, and field communication retrofits. Each controller shows both PV (process value) and SP (set value) readings.
Call our Applications Specialists for more information on these products or to special order a controller with other options.
Call For Price
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
DIN Size On / Off Function PID Function
Availability Pricing
West Controls 95001PA000 1/16-DIN Temp Controller, 2-Line; Sensor/SSRD/2 Relay WZ-89820-71 Mfr # 95001PA000
COMPARE
1/16
Call For Price
West Controls 95001PA200 1/16-DIN Temp Controller, 2-Line; Sensor/SSRD/2 Relay/RS-232 WZ-89820-72 Mfr # 95001PA200
COMPARE
1/16
Call For Price
West Controls 95001PA400 1/16-DIN Temp Controller, 2-Line; Sensor/SSRD/2 Relay/RS-485 WZ-89820-73 Mfr # 95001PA400
COMPARE
1/16
Call For Price
West Controls 95B11PA000 1/16-DIN Temp Controller, 2-Line; Sensor/4-20mA/2 Relay WZ-89820-74 Mfr # 95B11PA000
COMPARE
1/16
Call For Price
West Controls 95B11PB000 1/16-DIN Temp Controller, 2-Line; 4-20mA/4-20mA/2 Relay WZ-89820-75 Mfr # 95B11PB000
COMPARE
1/16