Burrell Mighty-Magnum Wrist Action Shaker
Variable speed from a gentle rocking motion to violent agitation
  • Solid weight and design prevents creeping, crawling, or walking during use
  • Easily shakes loads up to 30 lb (13.6 kg)
Heavy-duty 1/8 hp motor allows the user to shake flasks, bottles, or jars up to 1000-mL. Featuring a fixed 10° arc and adjustable oscillations up to 600 per minute, the shaker can be operated in continuous mode or timed from 1 sec to 99 min 59 seconds. Bright LED screen clearly displays oscillations.
£10,448.97GBP / Each
£12,538.76GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Capacity
Availability Pricing
Burrell 075-790-08-45 Mighty Magnum Wrist Action Shaker, 8 Vessels, 100 - 240 VAC. WZ-51100-04 Mfr # 075-790-08-45
COMPARE
4 Vessels
£10,448.97GBP / Each
£12,538.76GBP (incl VAT)