This product is now discontinued, View Related Products
BriskHeat® Briskheat Heavy Insulated Samox® Fiberglass High Temperature Heating Tape with X2 PID Temperature Controller Bundles
Ready-to-use heating cord bundles deliver complete systems for exceptional convenience
  • Programmable controller features alarm, user selectable °C or °F, and advanced PID control
  • Easy to read measurements with 0.4” high, 4-digit, green LED display
  • Type J thermocouple provides feedback for strict temperature control
  • Enhanced durability with heavy insulated design
  • Constructed of heavy insulated knitted and braided Somax® fiberglass
All-inclusive bundle features a digital PID controller with type J thermocouple, heating tape, and power cords—everything you need to get started. Benchtop controller is compact and easy to program, featuring a plug-and-play design. The X2 series controller offers a simple three button control with a programmable temperature range of 0° to 760°C (32° to 1400°F). This advanced controller offers an accuracy with Type-J heating cords of ±0.5°C (±0.9°F).
The heavy insulated heating tapes feature an IP5X, rating making them ideal for rugged applications. Select heating tape length to optimize coverage while avoiding overlap.
Call For Price
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Length (ft) Width (in) Description
Availability Pricing
BriskHeat X2J-BWH051020L Heavy Insulated Samox® Fiberglass High Temperature Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 0.5 in x 2.0 ft; 120 VAC WZ-36120-52 Mfr # X2J-BWH051020L
COMPARE
2 1/2 Heavy Insulated Samox® Fiberglass High Temperature Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 0.5 in x 2.0 ft; 120 VAC
Call For Price
BriskHeat X2J-BWH051040L Heavy Insulated Samox® Fiberglass High Temperature Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 0.5 in x 4.0 ft; 120 VAC WZ-36120-53 Mfr # X2J-BWH051040L
COMPARE
4 1/2 Heavy Insulated Samox® Fiberglass High Temperature Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 0.5 in x 4.0 ft; 120 VAC
Call For Price
BriskHeat X2J-BWH051060L Heavy Insulated Samox® Fiberglass High Temperature Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 0.5 in x 6.0 ft; 120 VAC WZ-36120-54 Mfr # X2J-BWH051060L
COMPARE
6 1/2 Heavy Insulated Samox® Fiberglass High Temperature Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 0.5 in x 6.0 ft; 120 VAC
Call For Price
BriskHeat X2J-BWH051080L Heavy Insulated Samox® Fiberglass High Temperature Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 0.5 in x 8.0 ft; 120 VAC WZ-36120-55 Mfr # X2J-BWH051080L
COMPARE
8 1/2 Heavy Insulated Samox® Fiberglass High Temperature Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 0.5 in x 8.0 ft; 120 VAC
Call For Price
BriskHeat X2J-BWH101020L Heavy Insulated Samox® Fiberglass High Temperature Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 1.0 in x 2.0 ft; 120 VAC WZ-36120-56 Mfr # X2J-BWH101020L
COMPARE
2 1 Heavy Insulated Samox® Fiberglass High Temperature Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 1.0 in x 2.0 ft; 120 VAC
Call For Price
BriskHeat X2J-BWH101040L Heavy Insulated Samox® Fiberglass High Temperature Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 1.0 in x 4.0 ft; 120 VAC WZ-36120-57 Mfr # X2J-BWH101040L
COMPARE
4 1 Heavy Insulated Samox® Fiberglass High Temperature Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 1.0 in x 4.0 ft; 120 VAC
Call For Price
BriskHeat X2J-BWH101060L Heavy Insulated Samox® Fiberglass High Temperature Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 1.0 in x 6.0 ft; 120 VAC WZ-36120-58 Mfr # X2J-BWH101060L
COMPARE
6 1 Heavy Insulated Samox® Fiberglass High Temperature Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 1.0 in x 6.0 ft; 120 VAC
Call For Price
Showing 1- 7 of 7