BRAND® Transferpette® Multi-Channel Electronic Pipettors
Intuitive operation in an "Ergonomics Approved" pipette!
  • Microprocessor control for precise volume delivery
  • Choose from five convenient operating modes to improve productivity
  • “Gel” mode discharges slowly for electrophoresis gels
  • Eliminate user variability
  • Easy maintenance
These multi-channel electronic pipettors feature a soft-touch pipetting key, contoured handle, and an adjustable finger rest for maximum comfort during use. The prestigious TUV testing service, an OSHA-accredited independent testing lab, has given these electronic pipettors the “Ergonomic Approved” certification.
Large LCD screen clearly displays all settings. Each function has a dedicated control to reduce inadvertent volume changes or tip ejection. Rechargeable battery provides over 4000 pipetting cycles on a singe charge. Units can be disassembled for cleaning and pipette shaft may be removed and autoclaved for decontamination or use in sterile applications. Easy Calibration™ system reduces the need for off-site calibration and simplifies conformance with ISO/GLP guidelines.
Operating modes:
Pipetting mode aspirates and discharges a preset volume.
Mixing mode repeatedly aspirates and discharges a preset volume and display shows number of completed cycles.
Reverse mode is designed for high viscosity, high vapor pressure, and low surface tension liquids.
Dispensing mode discharges a preset number of small equal aliquots from the total aspirated volume.
Gel mode Special mode for loading of electrophoresis gels.
£1,349.16GBP / Each
£1,618.99GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (mL) Min Volume (mL) Number Of Channels
Availability Pricing
BRAND Transferpette® Pipettor Electronic 12 Channel, 1-20ul per Channel; 110 V Charger WZ-25016-32 Mfr # 705470
COMPARE
0.02 0.001 12
£1,349.16GBP / Each
£1,618.99GBP (incl VAT)
BRAND Transferpette® Pipettor Electronic 12 Channel, 10-200ul per Channel; 110 V Charger WZ-25016-36 Mfr # 705474
COMPARE
0.2 0.01 12
£1,349.16GBP / Each
£1,618.99GBP (incl VAT)