BRAND® BRANDplates® cellGrade™, cellGrade™ plus, cellGrade™ premium and inertgrade™ Cell Culture Microplates
Minimizes cross-contamination and ensures maximize results
  • cellGrade™ standard cell culture treatment
  • cellGrade™ plus enhanced surface for more fastidious cells, or reduced serum culture
  • cellGrade™ preminum poly-D lysine equivalent for serum-free culture, or the most fastidious cells
  • inertGrade for suspension cultivation to inhibit differentiation of stem cells
  • Sterility Assurance Level (SAL) of 10-6
These cell culture microplates come in a variety of surface treatments ideal for use in the culture of a wide range of cell types. The plates are sterilized by ß-radiation or ethylene oxide, are DNA-, DNase-, RNase-free, and are packaged individually wrapped.
The polystyrene microplates are designed to lessen cross contamination, ensure outstanding recovery, and offer many options with color and number of wells. Three different bottom shapes are available: round (U) with no edges, flat (F) with a flat bottom, and flat with curved edges for enhanced mixing (C) bottom. For easy identification of the surface type and well location, the 96-well standard format plates feature an orange color-coded embossed alphanumeric lettering for easy identification of wells.
With a chimney well type of design, each well is separated from each other to reduce the risk of cross contamination. The 384-well microplates feature rounded corners which allow optimal uniform meniscus formation.
Convenient accessories for use with the microplates, such as lids, rubber rollers, and sealing films are offered.
£324.78 - £1,228.31GBP / Pkg of 50
£389.74 - £1,473.97GBP (incl VAT)
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Number of Wells Surface Treatment Well Shape
Availability Pricing
BRANDplates® cellGrade™  Sterile Microplate, 96-Well, PS, White, 350 µL, Standard F-Bottom; 50/PK WZ-07903-59 Mfr # 781965
COMPARE
96 cellGrade™ Standard F-Bottom
£324.78GBP / Pkg of 50
£389.74GBP (incl VAT)
BRANDplates® cellGrade™  Sterile Microplate, 96-Well, PS, Black, 350 µL, Standard F-Bottom; 50/PK WZ-07903-61 Mfr # 781968
COMPARE
96 cellGrade™ Standard F-Bottom
£324.78GBP / Pkg of 50
£389.74GBP (incl VAT)
BRANDplates® cellGrade™  Sterile Microplate, 96-Well, PS, Black, 330 µL, Clear F-Bottom; 50/PK WZ-07903-63 Mfr # 781971
COMPARE
96 cellGrade™ Clear F-Bottom
£794.61GBP / Pkg of 50
£953.53GBP (incl VAT)
BRANDplates® cellGrade™  Sterile Microplate, 384-Well, PS, Clear, 100 µL, Standard F-Bottom; 50/PK WZ-07903-64 Mfr # 781980
COMPARE
384 cellGrade™ Standard F-Bottom
£506.02GBP / Pkg of 50
£607.22GBP (incl VAT)
BRANDplates® cellGrade™  Sterile Microplate, 384-Well, PS, White, 120 µL, Clear F-Bottom; 50/PK WZ-07903-68 Mfr # 781986
COMPARE
384 cellGrade™ Clear F-Bottom
£1,023.37GBP / Pkg of 50
£1,228.04GBP (incl VAT)
BRANDplates® cellGrade™ Premium Sterile Microplate, 96-Well, PS, White, 330 µL, Clear F-Bottom; 50/PK WZ-07903-81 Mfr # 782090
COMPARE
96 cellGrade™ premium Clear F-Bottom
£1,228.31GBP / Pkg of 50
£1,473.97GBP (incl VAT)