BRAND® Tips for Transferpettor™ Positive Displacement Pipettes
Color coded to match appropriate sized pipettes
  • Robust polypropylene (PP) construction
  • Reusable
Reusable tips are specially designed for the Transferpettor positive displacement pipettor.
£37.10 - £47.51GBP / Pkg of 10
£44.52 - £57.01GBP (incl VAT)
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Code Max Volume (mL) Min Volume (mL)
Availability Pricing
BRAND Transferpettor 702852 Positive Displacement Pipettor Tips, PP, 100-500 µL, 10/PK WZ-24550-75 Mfr # 702852
COMPARE
Green 0.5 0.1
£37.10GBP / Pkg of 10
£44.52GBP (incl VAT)
BRAND Transferpettor 702854 Positive Displacement Pipettor Tips, PP, 200-1000 µL, 10/PK WZ-24550-76 Mfr # 702854
COMPARE
Yellow 1 0.2
£47.51GBP / Pkg of 10
£57.01GBP (incl VAT)
BRAND Transferpettor 702858 Positive Displacement Pipettor Tips, PP, 1-5 mL, 10/PK WZ-24550-77 Mfr # 702858
COMPARE
Red 5 1
£37.10GBP / Pkg of 10
£44.52GBP (incl VAT)
BRAND Transferpettor 702860 Positive Displacement Pipettor Tips, PP, 2-10 mL, 10/PK WZ-24550-78 Mfr # 702860
COMPARE
Orange 10 2
£47.51GBP / Pkg of 10
£57.01GBP (incl VAT)