BRAND® Microcentrifuge Tube Racks
Keep your workbench samples organized with microcentifuge tube racks
  • For 1.5 mL tubes
  • Choice of 20- ,84- and 128-tube racks
  • Made of autoclavable polypropylene
Whether you're processing samples or transporting them, these BRAND® microcentrifuge tube racks are a laboratory essential. The racks are numbered for easy placement of tubes. The racks that hold 84 and 128 tubes are stackable, too.
£18.10 - £92.34GBP / Pkg of 5
£21.72 - £110.81GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Max Tube Size (mm) Tubes Held
Availability Pricing
BRAND Microcentrifuge, Stackable 84-Tube Rack, Polypropylene, Blue, up to 13 mm Tube dia.; 5/PK WZ-06101-24 Mfr # 4341001
COMPARE
Blue 13 84
£92.34GBP / Pkg of 5
£110.81GBP (incl VAT)
BRAND Microcentrifuge, Stackable 128-Tube Rack, Polypropylene, Blue, up to 11 mm Tube dia.; 5/PK WZ-06101-28 Mfr # 4341051
COMPARE
Blue 11 128
£18.10GBP / Pkg of 5
£21.72GBP (incl VAT)