Bradley HALO Eye/Face Wash Bowls
No struggling to activate the eyewash – broad hand flag makes activation easy!
  • Eye/Face wash bowls provide excellent face wash down coverage
HALO eyewashes provide rapid relief to individuals who have been injured by chemicals or particulates. Ergonomic hand activation is easy to reach from both sides of the bowl. Integral strainer reduces debris in the supply water for further eye protection. Bowls are contoured for superior draining, reducing the risk of bacterial growth when not in use. Separate supply and waste pipes meet CEN and plumbing requirements. Supply connection is 1/2" NPT(F); waste connection is 1-1/4" NPT(M).
Compliance: ANSI Z358.1-2009
£458.82GBP / Each
£550.58GBP (incl VAT)
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Bowl Material Mount Description
Availability Pricing
Bradley S19224PDCFW Eye/Face wash, Wall mount, ABS plastic with cover WZ-86001-32 Mfr # S19224PDCFW
COMPARE
ABS plastic Wall Mount Eye/Face wash, Wall mount, ABS plastic with cover
£458.82GBP / Each
£550.58GBP (incl VAT)