This product is now discontinued, View Related Products

Bradley Combination Showers

Stay-open valves and flow control ensure constant rinsing flow.
All units feature plastic shower heads with either yellow ABS plastic, corrosive-resistant PVC, or stainless steel eyewash/facewash bowls (10" dia). Each shower comes with safety sign and inspection tag.
Call For Price

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Supply Pipe Material Description
Availability Pricing
Bradley S19-310 Combination Showers-Shower/eyewash station WZ-86000-40 Mfr # S19-310
COMPARE
Galvanized steel Combination Showers-Shower/eyewash station
Call For Price
Bradley S19-310FW Combination Showers-Shower/eye/facewash station WZ-86000-50 Mfr # S19-310FW
COMPARE
Galvanized steel Combination Showers-Shower/eye/facewash station
Call For Price
Bradley S19-310AC Combination Showers (foot operated) WZ-86000-52 Mfr # S19-310AC
COMPARE
Galvanized steel Combination Showers (foot operated)
Call For Price
Bradley S19-310PVC Combination Showers-plastic contruction WZ-86000-54 Mfr # S19-310PVC
COMPARE
PVC Combination Showers-plastic contruction
Call For Price
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat