This product is now discontinued, View Related Products
Body Filter 95+ Coveralls 63440
Fabric retains >95% of 0.3 micron particulates while allowing high air flow for breathability
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
  • Protects against dangerous noxious particulates
These coveralls are constructed of electrostatically-charged filter media for superior protection against particulates. Polymer grid on the exterior surface provides exceptional fabric strength and abrasion resistance. The smooth interior contact surface wicks moisture away from the body for added comfort. The highly-efficient filtration media also provides greater air permeability of 8.65 cfm at 20 Pa, as tested in accordance with ASTM D737.
Call For Price
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
Compareitem specifications
Size Description
Availability Pricing
International Enviroguard Coveralls, elastic wrists, medium, 25/cs SKU 8622625 WZ-86226-25 Mfr # 4012-M
COMPARE
Medium Coveralls, elastic wrists, medium, 25/cs
Call For Price
International Enviroguard Coveralls, elastic wrists, large, 25/cs SKU 8622626 WZ-86226-26 Mfr # 4012-L
COMPARE
Large Coveralls, elastic wrists, large, 25/cs
Call For Price
International Enviroguard Coveralls, elastic wrists, extra large, 25/cs SKU 8622627 WZ-86226-27 Mfr # 4012-XL
COMPARE
X-large Coveralls, elastic wrists, extra large, 25/cs
Call For Price
International Enviroguard Coveralls, elastic wrists, double extra large, 25/cs SKU 8622628 WZ-86226-28 Mfr # 4012-2XL
COMPARE
XX-large Coveralls, elastic wrists, double extra large, 25/cs
Call For Price
International Enviroguard Coveralls, elastic wrists and ankles, medium, 25/cs WZ-86226-35 Mfr # 4017-M
COMPARE
Medium Coveralls, elastic wrists and ankles, medium, 25/cs
Call For Price
International Enviroguard Coveralls, elastic wrists and ankles, large, 25/cs WZ-86226-36 Mfr # 4017-L
COMPARE
Large Coveralls, elastic wrists and ankles, large, 25/cs
Call For Price
International Enviroguard Coveralls, elastic wrists and ankles, extra large, 25/cs WZ-86226-37 Mfr # 4017-XL
COMPARE
X-large Coveralls, elastic wrists and ankles, extra large, 25/cs
Call For Price
International Enviroguard Coveralls, elastic wrists and ankles, double extra large, 25/cs WZ-86226-38 Mfr # 4017-2XL
COMPARE
XX-large Coveralls, elastic wrists and ankles, double extra large, 25/cs
Call For Price
International Enviroguard Coveralls, with hood, elastic wrists, medium, 25/cs SKU 8622645 WZ-86226-45 Mfr # 4028-M
COMPARE
Medium Coveralls, with hood, elastic wrists, medium, 25/cs
Call For Price
International Enviroguard Coveralls, with hood, elastic wrists, large WZ-86226-46 Mfr # 4028-L
COMPARE
Large Coveralls, with hood, elastic wrists, large
Call For Price
International Enviroguard Coveralls, with hood, elastic wrists, extra large, 25/cs SKU 8622647 WZ-86226-47 Mfr # 4028-XL
COMPARE
X-large Coveralls, with hood, elastic wrists, extra large, 25/cs
Call For Price
International Enviroguard Coveralls, with hood, elastic wrists, double extra large, 25/cs WZ-86226-48 Mfr # 4028-2XL
COMPARE
XX-large Coveralls, with hood, elastic wrists, double extra large, 25/cs
Call For Price
International Enviroguard Coveralls, with hood and boots, elastic wrists, medium, 25/cs SKU 8622655 WZ-86226-55 Mfr # 4014-M
COMPARE
Medium Coveralls, with hood and boots, elastic wrists, medium, 25/cs
Call For Price
International Enviroguard Coveralls, with hood and boots, elastic wrists, large, 25/cs SKU 8622656 WZ-86226-56 Mfr # 4014-L
COMPARE
Large Coveralls, with hood and boots, elastic wrists, large, 25/cs
Call For Price
International Enviroguard Coveralls, Elastic Wrist, Hood/Boots, XL; 25/Cs WZ-86226-57 Mfr # 4014-XL
COMPARE
X-large Coveralls, Elastic Wrist, Hood/Boots, XL; 25/Cs
Call For Price
International Enviroguard Coveralls, Elastic Wrist, Hood/Boots, XXL; 25/Cs WZ-86226-58 Mfr # 4014-2XL
COMPARE
XX-large Coveralls, Elastic Wrist, Hood/Boots, XXL; 25/Cs
Call For Price
Showing 1- 16 of 16