Environmental Express® BOD Bottle Storage Racks and Carrier
Safe and convenient organization of BOD bottles
  • Stackable
  • Choice of polypropylene or steel construction
Polypropylene racks can be autoclaved up to 121°C. Steel racks are more durable but cannot be autoclaved.
£73.26 - £116.06GBP / Each
£87.91 - £139.27GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Length (in) Material Description
Availability Pricing
COMPARE
13 Epoxy-coated steel BOD Bottle Storage Rack, Epoxy-coated Steel, 300 mL, 12-place; Each
£73.26GBP / Each
£87.91GBP (incl VAT)
COMPARE
13 Epoxy-coated steel BOD Bottle Storage Rack, Epoxy-Coated Steel, 300 mL, 12-place
£116.06GBP / Pkg of 1
£139.27GBP (incl VAT)