BRAND® BLAUBRAND® USP Bulb Pipettes
Accurate measurement of specific volumes
  • High-quality AR-GLAS®
  • High-contrast blue enamel markings for legibility
  • Calibrated "To Deliver" (TD, Ex) to DIN EN ISO 648
These bulb pipettes comply with the Class A error limits required by the US Pharmacopeia. Each pipette is calibrated "to deliver" on computer controlled, automated production lines for maximum precision. All pipettes come with a batch number and an accompanying batch certificate.
£17.10 - £230.34GBP / Pkg of 6
£20.52 - £276.41GBP (incl VAT)
17 variations of this product are available.
See All Product Options Below
17 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Code Max Volume (mL) Overall Length (mm)
Availability Pricing
BRAND BLAUBRAND®  Bulb Pipette, Glass, Class A, 0.5 mL; 12/PK WZ-25210-18 Mfr # 30601
COMPARE
2x Black 0.5 300
£206.25GBP / Pkg of 12
£247.50GBP (incl VAT)
BRAND BLAUBRAND®  Bulb Pipette, Glass, Class A, 1 mL; 12/PK WZ-25210-19 Mfr # 30602
COMPARE
Blue 1 300
£207.30GBP / Pkg of 12
£248.76GBP (incl VAT)
BRAND BLAUBRAND®  Bulb Pipette, Glass, Class A, 2 mL; 12/PK WZ-25210-20 Mfr # 30603
COMPARE
Orange 2 330
£230.34GBP / Pkg of 12
£276.41GBP (incl VAT)
BRAND BLAUBRAND®  Bulb Pipette, Glass, Class A, 3 mL; 6/PK WZ-25210-21 Mfr # 30605
COMPARE
Black 3 330
£55.61GBP / Pkg of 6
£66.73GBP (incl VAT)
BRAND BLAUBRAND®  Bulb Pipette, Glass, Class A, 4 mL; 6/PK WZ-25210-22 Mfr # 30606
COMPARE
2x red 4 400
£64.42GBP / Pkg of 6
£77.30GBP (incl VAT)
BRAND BLAUBRAND®  Bulb Pipette, Glass, Class A, 5 mL; 6/PK WZ-25210-23 Mfr # 30607
COMPARE
White 5 400
£143.96GBP / Pkg of 6
£172.75GBP (incl VAT)
BRAND BLAUBRAND®  Bulb Pipette, Glass, Class A, 6 mL; 6/PK WZ-25210-24 Mfr # 30608
COMPARE
2x Orange 6 400
£76.15GBP / Pkg of 6
£91.38GBP (incl VAT)
BRAND BLAUBRAND®  Bulb Pipette, Glass, Class A, 8 mL; 6/PK WZ-25210-25 Mfr # 30610
COMPARE
Blue 8 440
£76.15GBP / Pkg of 6
£91.38GBP (incl VAT)
BRAND BLAUBRAND®  Bulb Pipette, Glass, Class A, 9 mL; 6/PK WZ-25210-26 Mfr # 30611
COMPARE
Black 9 440
£76.15GBP / Pkg of 6
£91.38GBP (incl VAT)
BRAND BLAUBRAND®  Bulb Pipette, Glass, Class A, 10 mL; 6/PK WZ-25210-27 Mfr # 30612
COMPARE
Red 10 440
£57.29GBP / Pkg of 6
£68.75GBP (incl VAT)
BRAND BLAUBRAND®  Bulb Pipette, Glass, Class A, 15 mL; 6/PK WZ-25210-28 Mfr # 30613
COMPARE
Green 15 510
£70.70GBP / Pkg of 6
£84.84GBP (incl VAT)
BRAND BLAUBRAND®  Bulb Pipette, Glass, Class A, 20 mL; 6/PK WZ-25210-29 Mfr # 30614
COMPARE
Yellow 20 510
£71.40GBP / Pkg of 6
£85.68GBP (incl VAT)
BRAND BLAUBRAND®  Bulb Pipette, Glass, Class A, 25 mL; 6/PK WZ-25210-30 Mfr # 30615
COMPARE
Blue 25 520
£198.66GBP / Pkg of 6
£238.39GBP (incl VAT)
BRAND BLAUBRAND®  Bulb Pipette, Glass, Class A, 30 mL; 6/PK WZ-25210-31 Mfr # 30616
COMPARE
Black 30 520
£84.96GBP / Pkg of 6
£101.95GBP (incl VAT)
BRAND BLAUBRAND®  Bulb Pipette, Glass, Class A, 40 mL; 6/PK WZ-25210-32 Mfr # 30617
COMPARE
White 40 540
£17.10GBP / Pkg of 6
£20.52GBP (incl VAT)
BRAND BLAUBRAND®  Bulb Pipette, Glass, Class A, 50 mL; 6/PK WZ-25210-33 Mfr # 30618
COMPARE
Red 50 540
£96.27GBP / Pkg of 6
£115.52GBP (incl VAT)
BRAND BLAUBRAND®  Bulb Pipette, Glass, Class A, 100 mL; 6/PK WZ-25210-34 Mfr # 30619
COMPARE
Yellow 100 585
Call For Price
Showing 1- 17 of 17