BIOLiner High Capacity Fixed Angle Rotor, Buckets, and Adapters
High capacity rotor provides versatility and optional biocontainment capabilities
The BIOLiner high capacity rotor provides versatility to your system and provides protection from samples with the optional biocontainment lids. Lids snap easily in place with just one hand. Three easy steps to change your configuration:
  • Release rotor by pressing button located in the center of the rotor.
  • Remove and attach new buckets/adapters by snapping into place.
  • Press rotor back in place until it is secure.
Adapters provide you additional versatility by accomodating sample containers from 5 mL collection tubes to 750 mL flat bottom bottles. Choose the adapters and buckets that meet your needs.
£569.90 - £2,170.97GBP / Each
£683.88 - £2,605.16GBP (incl VAT)
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Thermo Scientific 75003667 BIOLiner Swinging-Bucket Rotor w/o Buckets/Adapters/Lids WZ-17708-40 Mfr # 75003667
COMPARE
BIOLiner Swinging-Bucket Rotor w/o Buckets/Adapters/Lids
£2,170.97GBP / Each
£2,605.16GBP (incl VAT)
Thermo Scientific 75003668 Buckets For Bioliner Rotor 17708-40, 4/pk WZ-17708-41 Mfr # 75003668
COMPARE
Buckets For Bioliner Rotor 17708-40, 4/pk
£1,938.44GBP / Pkg of 4
£2,326.13GBP (incl VAT)
Thermo Scientific 75003669 Sealing Lids For Bioliner Buckets 17708-41, 4/pk WZ-17708-42 Mfr # 75003669
COMPARE
Sealing Lids For Bioliner Buckets 17708-41, 4/pk
£714.06GBP / Pkg of 4
£856.87GBP (incl VAT)
Thermo Scientific 75003675 Centrifuge Adapters, Oak Ridge 250 mL for BIOLiner; 4/Pk WZ-17708-46 Mfr # 75007305
COMPARE
Centrifuge Adapters, Oak Ridge 250 mL for BIOLiner; 4/Pk
£569.90GBP / Pkg of 4
£683.88GBP (incl VAT)
Thermo Scientific 75003674 Adapters For 50 mL Conical Tubes (10 Positions), Fits Bucket 17708-41, 4/pk WZ-17708-47 Mfr # 75003674
COMPARE
Adapters For 50 mL Conical Tubes (10 Positions), Fits Bucket 17708-41, 4/pk
£786.08GBP / Pkg of 4
£943.30GBP (incl VAT)
Thermo Scientific 75003671 Centrifuge Adapters, 5-7 mL for BIOLiner; 4/Pk WZ-17708-51 Mfr # 75003671
COMPARE
Centrifuge Adapters, 5-7 mL for BIOLiner; 4/Pk
£636.89GBP / Pkg of 4
£764.27GBP (incl VAT)