This product is now discontinued, View Related Products
Binder VDL Standard Series Vacuum Ovens for Flammable Solvents
For complete safety during drying of organic solvents, these chambers bear the GS marking
  • VDL series ovens can dry explosive substances containing solvents
  • Gentle and quick drying
  • Thermal conducting plates provide direct heat transfer
  • Has a proven safety concept and ATEX-certified interior
These drying chambers operate quickly and are free of residues, scaling, or oxidation. The chamber can adjust without overshooting to protect sensitive samples. Designed for flammable solvents, the interior is explosion-proof. All equipment complies with ATEX II 2/3/- G standards, guaranteeing operator safety. The pressure-encapsulated instrument box, preheating chamber, and heating elements use compressed air to prevent solvent vapors from igniting in the electric zone.
Conductor plates transfer heat directly to the sample, while the APT.line™ preheating chamber technology promises a homogeneous drying across the entire usable space. Drying happens on adjustable aluminum expansion racks that are free form condensation. The temperature range of 8 °C to 110 °C offers a high level of process reliability. The microprocessor controller with LED display allows easy programming of individual temperature profiles and advanced applications for reproducible drying processes. Calibration can be achieved with the logical function keys. All ovens have an RS-232 port, inert gas connection with fine-dosing aeration valve, analog manometer, and integral measuring gauge. The integral thermostat is rated Class 2, according to DIN 12880, Part 1.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem Availability Pricing
Binder VDL023-230V Standard Series Vacuum Drying Chambers, for Flammable Solvents, 23 L; 230 VAC, 50/60 Hz WZ-05012-96 Mfr # 9030-0038
COMPARE
Call For Price
Binder VDL053-230V Standard Series Vacuum Drying Chambers, for Flammable Solvents, 53 L; 230 VAC, 50/60 Hz WZ-05012-97 Mfr # 9030-0039
COMPARE
Call For Price
Binder VDL115-230V Standard Series Vacuum Drying Chambers, for Flammable Solvents, 115 L; 230 VAC, 50/60 Hz WZ-05012-98 Mfr # 9030-0040