Cole-Parmer® Beaker Positioning Covers for Ultrasonic Cleaners
Achieve optimal cleaning results
  • Ultrasonic activity is greatest about an inch from the bottom of the tank, so suspending your items provides the best cleaning possible and helps prevent damage to the transducers
Use polypropylene beaker positioning covers with beakers to clean small objects or for indirect cleaning. Cover suspends beakers above tank bottom for increased ultrasonic activity. Covers come with beaker suspension ring. Order beakers separately.
£58.49 - £102.79GBP / Each
£70.19 - £123.35GBP (incl VAT)
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Material Description
Availability Pricing
COMPARE
305 SS Beaker Positioning Cover for 0.5 gal and SS 0.75 gal Cleaners
£58.49GBP / Each
£70.19GBP (incl VAT)
COMPARE
304 SS Beaker Positioning Cover for 0.75 gal and SS 1 gal Cleaners
£61.98GBP / Each
£74.38GBP (incl VAT)
COMPARE
307 SS Beaker Positioning Cover for 1.5 gal and SS 1.5 gal Cleaners
£80.62GBP / Each
£96.74GBP (incl VAT)
COMPARE
306 SS Beaker Positioning Cover for 2.5 gal and SS 2.5/3.5 gal Cleaners
£87.68GBP / Each
£105.22GBP (incl VAT)
COMPARE
308 SS Beaker Positioning Cover for 5.5 gal and SS 5.5 gal Cleaners
£102.79GBP / Each
£123.35GBP (incl VAT)