Analytik Jena Battery-Operated Ultraviolet Lamps
Portable design is great for use in the field
  • Mini built-in flashlight is included
Choose from single- or dual-wavelength lamps; all include 6-watt tube(s). Single wavelength lamps let you use both tubes simultaneously to achieve intensities twice as high as similar units. Dual-wavelength lamps let you choose from either 254 or 365 nm light.
Battery-operated ultraviolet lamps are made of lightweight CYCOLAC(R) for easy, fatigue-free use. All models have a built-in flashlight; order replacement white bulbs from "Accessories." Powered by two 6V lantern batteries (sold separately); average battery life is 20 hrs. Lamps measure 2-7/8"W x 9-1/2"H x 9-3/8"D. Shpg wt 2.6 lb (1.2 kg).
£529.48GBP / Each
£635.38GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Intensity (µW/cm2) Width (in)
Availability Pricing
Analytik Jena Battery-operated UV lamp, 254 nm wavelength WZ-09817-60 Mfr # 95-0009-02
COMPARE
1350 2 7/8
£529.48GBP / Each
£635.38GBP (incl VAT)
Analytik Jena Battery-operated UV lamp, 365 nm wavelength WZ-09817-80 Mfr # 95-0011-01
COMPARE
1650 2 7/8