Bag Filters for Polypropylene Bag Filter Housings
Choose from standard felt bags, high-efficiency bags, or nylon mesh bags. Each bag measures 4" diameter. The 10"L bags have a surface area of 0.75 ft2; 18-½"L bags have a surface area of 1.75 ft2.
£4.81 - £20.23GBP / Each
£5.77 - £24.28GBP (incl VAT)
34 variations of this product are available.
See All Product Options Below
34 PRODUCT OPTIONS
Compareitem specifications
Filter Length (in) Filter Media Micron Rating
Availability Pricing
COMPARE
10 Polypropylene 1
£6.74GBP / Each
£8.09GBP (incl VAT)
COMPARE
10 Polypropylene 10
£5.02GBP / Each
£6.02GBP (incl VAT)
COMPARE
10 Polypropylene 25
£4.81GBP / Each
£5.77GBP (incl VAT)
COMPARE
10 Polypropylene 50
£4.81GBP / Each
£5.77GBP (incl VAT)
COMPARE
10 Polypropylene 100
£4.81GBP / Each
£5.77GBP (incl VAT)
COMPARE
10 Polypropylene 200
£4.81GBP / Each
£5.77GBP (incl VAT)
COMPARE
18.5 Polypropylene 5
£6.11GBP / Each
£7.33GBP (incl VAT)
COMPARE
18.5 Polypropylene 10
£6.11GBP / Each
£7.33GBP (incl VAT)
COMPARE
18.5 Polypropylene 200
£6.11GBP / Each
£7.33GBP (incl VAT)
COMPARE
10 Polypropylene 25
£6.53GBP / Each
£7.84GBP (incl VAT)
COMPARE
10 Polypropylene 50
£8.05GBP / Each
£9.66GBP (incl VAT)
COMPARE
10 Nylon 200
£14.96GBP / Each
£17.95GBP (incl VAT)
COMPARE
10 Nylon 400
£11.80GBP / Each
£14.16GBP (incl VAT)
COMPARE
10 Nylon 800
£11.80GBP / Each
£14.16GBP (incl VAT)
COMPARE
18.5 Polypropylene 25
£16.44GBP / Each
£19.73GBP (incl VAT)
COMPARE
18.5 Polypropylene 50
£13.49GBP / Each
£16.19GBP (incl VAT)
COMPARE
18.5 Polypropylene 100
£13.49GBP / Each
£16.19GBP (incl VAT)
COMPARE
18.5 Nylon 100
£14.33GBP / Each
£17.20GBP (incl VAT)
COMPARE
18.5 Nylon 200
£14.33GBP / Each
£17.20GBP (incl VAT)
COMPARE
18.5 Nylon 800
£13.91GBP / Each
£16.69GBP (incl VAT)
Showing 1 - 20 of 34