This product is now discontinued, View Related Products
Bürkle Stainless Steel Samplers for Bulk Goods
Easily sample powders or granules at targeted depths
  • Constructed of 316L stainless steel
  • Quickly empty the sample without having to disassemble the sampler
  • Autoclavable up to 121°C for 20 minutes
These powder and granulate samplers are designed to take a target point sample at the position within the sample material that has precisely been determined. The sampler is closed when it is inserted into the sample material, opened when it reaches the target point, and closed again once the sample is obtained.
The stainless steel microsamplers without heads, models 06041-06 through 06041-13, are ideal for sampling small quantities of powder. These samplers will accept interchangeable heads, allowing you to sample different volumes. The 12 mm diameter samplers will accept heads capable of sampling volumes of 0.2, 0.5, or 1 mL. The 25 mm diameter samplers will accept heads that can sample volumes of 2, 5, or 10 mL. Samplers and heads are sold separately.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Diameter (in) Length (in) Description
Availability Pricing
Burkle 5307-1055 Small powder sampling lance tube, manual pointed plunger, 55 cm x 12 mm WZ-06041-06 Mfr # 5307-1055
COMPARE
0.47 21.7 Sampling Lance Tube, manual pointed plunger; 55 cm x 12 mm
Call For Price
Burkle 5307-1085 Small powder sampling lance tube, manual pointed plunger, 85 cm x 12 mm WZ-06041-07 Mfr # 5307-1085
COMPARE
0.47 33.5 Sampling Lance Tube, manual pointed plunger; 85 cm x 12 mm
Call For Price
Burkle 5307-1120 Small powder sampling lance tube, manual pointed plunger, 120 cm x 12 mm WZ-06041-08 Mfr # 5307-1120
COMPARE
0.47 47.2 Sampling Lance Tube, manual pointed plunger; 120 cm x 12 mm
Call For Price
Burkle 5307-2055 Small powder sampling lance tube, manual pointed plunger, 55 cm x 25 mm WZ-06041-09 Mfr # 5307-2055
COMPARE
1 21.7 Sampling Lance Tube, manual pointed plunger; 55 cm x 25 mm
Call For Price
Burkle 5307-2085 Small powder sampling lance tube, manual pointed plunger, 85 cm x 25 mm WZ-06041-11 Mfr # 5307-2085
COMPARE
1 33.5 Sampling Lance Tube, manual pointed plunger; 85 cm x 25 mm
Call For Price
Burkle 5307-2120 Small powder sampling lance tube, manual pointed plunger, 120 cm x 25 mm WZ-06041-13 Mfr # 5307-2120
COMPARE
1 47.2 Sampling Lance Tube, manual pointed plunger; 120 cm x 25 mm