Bürkle Large Aluminum Samplers for Powders and Granulates
Reliably sample various bulk goods, powders, and granulates for different sampling tasks
  • Efficient sampling made possible with the smallest gap possible between the inner and outer tube
  • Constructed of ultrapure materials for a durable design
These maintenance-free samplers allow for collection from the finest powders to large grains. They are suitable for sampling from all layers of the container, providing an accurate representation of the contents. These samplers come in different lengths, with single or multiple chambers and openings, and made of different material options for all different sampling methods. The 55 cm long samplers are suitable for sacks; the 85 cm long samplers are ideal for barrels, tubs, and mixed containers; and the 150 cm long samplers are appropriate for big bags, silos, and tanks.
Large aluminum samplers are ideal for coarse grains and large-volume sampling. The aluminum samplers feature 1 chamber with either 3, 5, or 7 openings, and are suitable for sampling through all layers of a container. The aluminum and PTFE sampler models feature 3, 5, or 7 individual closed chambers. These chambers are separated by PTFE separators, making them ideal for point sampling at a predetermined depth. All samplers can be steam sterilized up to 250°F (121°C).
£619.74 - £1,344.58GBP / Each
£743.69 - £1,613.50GBP (incl VAT)
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Material Volume (mL) Description
Availability Pricing
Burkle 5319-1085 Large Sampling Lance, Aluminum, 3 openings, one chamber; 880 mL capacity WZ-06041-56 Mfr # 5319-1085
COMPARE
Aluminium 880 Large all layer sampling lance, aluminum, 3 openings, one chamber, 85 cm length, 710 mm insertion depth; 880 mL capacity
£619.74GBP / Each
£743.69GBP (incl VAT)
Burkle 5319-1150 Sampling Lance, Aluminum, 5 openings, one chamber; 1700 mL capacity WZ-06041-57 Mfr # 5319-1150
COMPARE
Aluminium 1700 Large all layer sampling lance, aluminum, 5 openings, one chamber, 150 cm length, 1355 mm insertion depth; 1700 mL capacity
£877.61GBP / Each
£1,053.13GBP (incl VAT)
Burkle 5319-1250 Large Sampling ance, aluminum, 7 openings, one chamber, 250 cm length, 2355 mm insertion depth; 2900 mL capacity WZ-06041-58 Mfr # 5319-1250
COMPARE
Aluminium 2900 Large all layer sampling lance, aluminum, 7 openings, one chamber, 250 cm length, 2355 mm insertion depth; 2900 mL capacity
£991.37GBP / Each
£1,189.64GBP (incl VAT)
Burkle 5319-2085 Large Sampling Lance, Aluminum, 3 individual chambers; 390 mL capacity WZ-06041-59 Mfr # 5319-2085
COMPARE
Aluminium and PTFE 390 Large all layer sampling lance, aluminum, 3 individual chambers, 85 cm length, 710 mm insertion depth; 390 mL capacity
£957.78GBP / Each
£1,149.34GBP (incl VAT)
Burkle 5319-2150 Large Sampling Lance, Aluminum, 5 individual chambers; 650 mL capacity WZ-06041-60 Mfr # 5319-2150
COMPARE
Aluminium and PTFE 650 Large all layer sampling lance, aluminum, 5 individual chambers, 150 cm length, 1355 mm insertion depth; 650 mL capacity
£1,125.72GBP / Each
£1,350.86GBP (incl VAT)
Burkle 5319-2250 Sampling Lance, Aluminum, 7 individual chambers; 910 mL capacity WZ-06041-61 Mfr # 5319-2250
COMPARE
Aluminium and PTFE 910 Large all layer sampling lance, aluminum, 7 individual chambers, 250 cmlength, 2355 mm insertion depth; 910 mL capacity
£1,344.58GBP / Each
£1,613.50GBP (incl VAT)