Bürkle Ice-Boring Samplers
Attach to power drills to obtain frozen samples
  • Obtain samples up to a 7-7/8" (20 cm) depth
  • Choose from 5/8" (1.6 cm) or 2" (5 cm) diameter versions
  • Easily remove specimens with the bore rod (included)
These 316L stainless steel boring samplers feature sharp teeth to rapidly cut into deep-frozen or semisolid samples. The vented-crown design prevents air entrapment and allows for easier sample retrieval. A bore rod is also included to aid in specimen removal from the sampler.
The drill-bit head fits into standard power drills with keyless chucks (power drill not included).
Autoclavable.
£419.17 - £729.04GBP / Each
£503.00 - £874.85GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Diameter (in) Length (in) Material
Availability Pricing
Burkle 5323-0200 Ice and frozen substance sampling borer, SS, 16 x 21 mm; 20 cm deep WZ-06041-65 Mfr # 5323-0200
COMPARE
0.625 7.75 Stainless steel
£419.17GBP / Each
£503.00GBP (incl VAT)
Burkle 5323-2010 Ice and frozen substance sampling borer, SS, 50 x 54 mm; 20 cm deep WZ-06041-66 Mfr # 5323-2010
COMPARE
2 7.75 Stainless steel
£729.04GBP / Each
£874.85GBP (incl VAT)