This product is now discontinued, View Related Products
Bürkle Disposable Closed-Tip Piercing Lance Samplers
Easily take samples directly from the vessel with the durable piercing tip
  • Closed- and open-tip options available for different sampling methods
  • Manufactured in a cleanroom ISO Class 7 environment and individually packaged
  • Certified FDA 21 CFR 5 compliant for food applications
These HDPE samplers are ideal for taking large and small samples of powder and granulate directly from the bag. Choose from either sterile or nonsterile configurations. The sterilized options are sterilized via gamma irradiation.
The closed-tip samplers are suitable for cross-section samples. Because the tip is closed, the sample material will not fall out of the lance and will deliver a specific volume.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Length (in) Sterile Volume (mL)
Availability Pricing
Burkle 5393-5540 Disposable Closed-tip Sampling Picker Lance, HDPE, 300 mm x 21 mm; 60 mL WZ-06043-37 Mfr # 5393-5540
COMPARE
11.8 No 60
Call For Price
Burkle 5393-5541 Disposable Closed-tip Sampling Picker Lance HDPE, 300 mm x 21 mm, Sterile; 60 mL WZ-06043-38 Mfr # 5393-5541
COMPARE
11.8 Yes 60
Call For Price
Burkle 5393-5550 Disposable Closed-tip Sampling Picker Lance, HDPE, 500 mm x 21 mm; 100 mL WZ-06043-39 Mfr # 5393-5550
COMPARE
19.7 No 100
Call For Price
Burkle 5393-5551 Disposable Closed-tip Sampling Picker Lance, HDPE, 500 mm x 21 mm, Sterile; 100 mL WZ-06043-41 Mfr # 5393-5551
COMPARE
19.7 Yes 100