This product is now discontinued, View Related Products

Bürkle DispoPipette Disposable Industrial Pipette Samplers

Quickly take all-layer and target point samples with one sampler
  • Efficiently take samples of low-viscosity liquids
  • Produced and packaged according to cleanroom ISO Class 7
  • Constructed of durable HDPE for robust applications
DispoPipette disposable industrial pipette samplers are ideal for taking samples of low-viscosity liquids from carboys, drums, or tanks. Obtain all-layer samples when the sampler is inserted into the liquids while open and target point samples when the sampler is inserted while closed. With two opening sizes (2.5 and 5.5 mm) for sampling liquids of different viscosities. Available in two volume options, 100 and 200 mL, as well as nonsterilized or presterilized via gamma irradiation.
Call For Price

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Length (in) Sterile Volume (mL)
Availability Pricing
Burkle 5393-5500 Disposable Sampling Pipette, HDPE, 500 mm L; 100 mL WZ-06043-28 Mfr # 5393-5500
COMPARE
19.7 No 100
Call For Price
Burkle 5393-5501 Disposable Sampling Pipette, HDPE, 500 mm L, Sterile; 100 mL WZ-06043-29 Mfr # 5393-5501
COMPARE
19.7 Yes 100
Call For Price
Burkle 5393-5531 Disposable Sampling Pipette, HDPE, 1000 L; 200 mL WZ-06043-35 Mfr # 5393-5531
COMPARE
39.4 No 200
Call For Price
Burkle 5393-5532 Disposable Sampling Pipette, HDPE, 1000 mm L, Sterile; 200 mL WZ-06043-36 Mfr # 5393-5532
COMPARE
39.4 Yes 200
Call For Price
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat