Bürkle Automatic Self-Zeroing Shatter-Resistant Burettes (Burets)
Prevent breakage with these splinter-proof burettes
  • Constructed of glass with a plastic sheathing to resist breakage and glass splintering
  • Precise discharge jet allows very fine drops
  • Easy-to-read burette (buret) with blue graduations
These automatic self-zeroing burettes (burets) feature with a glass burette coated in plastic which increases resistance to breakage. If breakage does occur, the glass will not splinter. These burettes are constructed of polyethylene, polypropylene, natural rubber, and borosilicate glass wetted materials. Precise dosing is achievable with the clearly marked graduations as well as the press button and micro-screw control.
£120.95 - £133.67GBP / Each
£145.14 - £160.40GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Tolerance Description
Availability Pricing
Burkle 9695-3015 Titrating burette with shatter protection; 15 mL WZ-06044-50 Mfr # 9695-3015
COMPARE
500 0.045 Titrating burette with shatter protection; 15 mL
£120.95GBP / Each
£145.14GBP (incl VAT)
Burkle 9695-3025 Titrating burette with shatter protection; 25 mL WZ-06044-51 Mfr # 9695-3025
COMPARE
1000 0.045 Titrating burette with shatter protection; 25 mL
£120.95GBP / Each
£145.14GBP (incl VAT)
Burkle 9695-3050 Titrating burette with shatter protection; 50 mL WZ-06044-52 Mfr # 9695-3050
COMPARE
1000 0.075 Titrating burette with shatter protection; 50 mL
£133.67GBP / Each
£160.40GBP (incl VAT)