This product is now discontinued, View Related Products
B&K Precision Synthesized In-Circuit LCS - ESR Meter
With a built-in direct test fixture, testing the lead components is easy
  • Measurement parameters: Z, L, C, DCR, ESR, D, Q, and F
  • Test conditions: 100Hz, 120Hz, 1kHz, 10kHz, 100KHz (model 886 only), 1 Vrms, 0.25 Vrms, 0.05Vrms
  • 0.5% basic accuracy
The Model 885 and 886 Synthesized In-Circuit LCR/ESR meters have a wide range of test frequencies up to 10kHz for model 885 and 100kHz for model 886, and has many measurement parameters including Z, L, C, DCR, ESR, D, Q, and F as well. Both are designed for component evaluation on the production line and fundamental impedance testing for bench-top applications. With a built-in direct test fixture, testing the lead components is easy. The optional 4-wire test clip can give a convenient connection to larger components and assemblies with the accuracy of 4-wire testing.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Resistance Accuracy Resistance Range Resistance Resolution