B&K Precision Handheld Battery Capacity Analyzers
Test and maintain sealed lead acid (SLA) batteries
  • Measure battery voltage and capacity
  • Powered by the battery under test—no need for an external power supply
  • Rugged housing with durable Kelvin clamps
These lightweight, handheld battery capacity analyzers provide a quick, accurate test of percent capacity and voltage in 12 V lead acid batteries. Easily portable and ideal for field and shop service, maintenance applications with automotive fleets, communication equipment, UPS manufacturing and maintenance, and backup lighting systems.
£313.00 - £418.00GBP / Each
£375.60 - £501.60GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
B&K Precision 600B Handheld Battery Capacity Analyzer; 12 V SLA WZ-20025-98 Mfr # 600B
COMPARE
Handheld Battery Capacity Analyzer; 12 V SLA
£313.00GBP / Each
£375.60GBP (incl VAT)
B&K Precision 601B  Handheld Battery Capacity Analyzer; 6 and 12 V SLA WZ-20025-99 Mfr # 601B
COMPARE
Handheld Battery Capacity Analyzer; 6 and 12 V SLA
£418.00GBP / Each
£501.60GBP (incl VAT)