Cole-Parmer® Essentials Automatic Self-Zeroing, Self-Supporting Burettes (Burets)
Prevent contamination from outside elements
  • Quickly repeat titration by presetting drop speed with screw clamp
  • Easy-to-read burette (buret) with blue graduations
These burettes (burets) prevent contamination of media by outside elements. Feature easy-to-read blue graduations against white background. Flexible vinyl fill tube connects burette top with vinyl bottle stopper and extends into bottle. Operate the unique push-button stopcock using thumb pressure for drop-by-drop delivery. Small screw clamp lets you preset drop speed for repeat titration.
£112.69 - £387.00GBP / Each
£135.23 - £464.40GBP (incl VAT)
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Description
Availability Pricing
COMPARE
15 0.1 Automatic Self-Zeroing and Supporting Burette, 15 mL; 1/Pk
£112.69GBP / Each
£135.23GBP (incl VAT)
COMPARE
25 0.1 Automatic Self-Zeroing and Supporting Burette, 25 mL; 1/Pk
£122.06GBP / Each
£146.47GBP (incl VAT)
COMPARE
50 0.1 Automatic Self-Zeroing and Supporting Burette, 50 mL; 1/Pk
£387.00GBP / Each
£464.40GBP (incl VAT)