Setra ASM High-Accuracy Pressure Transducers
ASIC electronics provide constant and accurate output in extreme temperatures
  • ±0.05% full-scale accuracy
  • Compact, NEMA 4X housing
  • Gauge and absolute ranges
  • 4 to 20 mA output
Setra's AccuSense ASM series of pressure transducers are perfect for high-accuracy pressure applications requiring robust construction. The grooved design and hydraphobic porous plug located at the top of the unit protects the recessed air vent from contaminants in industrial environments. The ASIC electronics provides constant and accurate output in temperature extremes. Compact transducer features a robust digital sensor for excellent stability and outstanding performance.
Order SecureCal™ calibrator 98075-95 to calibrate tansducers in the field — enables zero-tare and snap-span adjustment, factory calibration restore function, and two-line LCD/LED feedback.
Other configurations such as a 7/16”-20 SAE straight thread connection are available; contact an Application Specialist.
£1,367.28GBP / Each
£1,640.74GBP (incl VAT)
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy Max Pressure (psia) Output
Availability Pricing
Setra ASM Pressure Transmitter, 0-15 PSIA WZ-98075-04 Mfr # ASM1015PAM11B3A00
COMPARE
±0.05% full-scale 15 4 to 20 mA
£1,367.28GBP / Each
£1,640.74GBP (incl VAT)
Setra ASM Pressure Transmitter, 0-25 PSIA WZ-98075-08 Mfr # ASM1025PA2M11B3A00
COMPARE
±0.05% full-scale 25 4 to 20 mA
£1,367.28GBP / Each
£1,640.74GBP (incl VAT)