Ashcroft® Utility Pressure Gauges, 1.5" to 2.5" Dial Sizes
Ideal to measure water, hydraulic oil, or gas pressure
  • True zero indication identifies damaged gauges—increases safety and ensures accuracy
Gauges include brass socket, soft-soldered phosphor bronze Bourdon tube, painted steel case, and polycarbonate lens.
£10.82 - £37.07GBP / Each
£12.98 - £44.48GBP (incl VAT)
20 variations of this product are available.
See All Product Options Below
20 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy Dial Size (in) Description
Availability Pricing
Ashcroft Utility Gauge, 30 to 0" Hg, 1 1/2" Dial, Back Connection WZ-68002-00 Mfr # 15-W-1005-P-H-01B-VAC/0#
COMPARE
±3-2-3% full-scale (ANSI/ASME Grade B) 1.5 30 to 0\" Hg Utility Gauge, 1 1/2\" Dial, Back Connection
£27.80GBP / Each
£33.36GBP (incl VAT)
Ashcroft Utility Gauge, 0 to 15 psi, 1 1/2" Dial, Back Connection WZ-68002-01 Mfr # 15-W-1005-P-H-01B-15#
COMPARE
±3-2-3% full-scale (ANSI/ASME Grade B) 1.5 0 to 15 psi Utility Gauge, 1 1/2\" Dial, Back Connection
£14.73GBP / Each
£17.68GBP (incl VAT)
Ashcroft Utility Gauge, 0 to 30 psi, 1 1/2" Dial, Back Connection WZ-68002-02 Mfr # 15-W-1005-P-H-01B-30#
COMPARE
±3-2-3% full-scale (ANSI/ASME Grade B) 1.5 0 to 30 psi Utility Gauge, 1 1/2\" Dial, Back Connection
£29.72GBP / Each
£35.66GBP (incl VAT)
Ashcroft Utility Gauge, 0 to 60 psi, 1 1/2" Dial, Back Connection WZ-68002-03 Mfr # 15-W-1005-P-H-01B-60#
COMPARE
±3-2-3% full-scale (ANSI/ASME Grade B) 1.5 0 to 60 psi Utility Gauge, 1 1/2\" Dial, Back Connection
£10.82GBP / Each
£12.98GBP (incl VAT)
Ashcroft Utility Gauge, 0 to 100 psi, 1 1/2" Dial, Back Connection WZ-68002-04 Mfr # 15-W-1005-P-H-01B-100#
COMPARE
±3-2-3% full-scale (ANSI/ASME Grade B) 1.5 0 to 100 psi Utility Gauge, 1 1/2\" Dial, Back Connection
£10.82GBP / Each
£12.98GBP (incl VAT)
Ashcroft Utility Gauge, 0 to 300 psi, 1 1/2" Dial, Back Connection WZ-68002-07 Mfr # 15-W-1005-P-H-01B-300#
COMPARE
±3-2-3% full-scale (ANSI/ASME Grade B) 1.5 0 to 300 psi Utility Gauge, 1 1/2\" Dial, Back Connection
£10.82GBP / Each
£12.98GBP (incl VAT)
Ashcroft Utility Gauge, 30 to 0" Hg, 2" Dial, Bottom Connection WZ-68003-00 Mfr # 20-W-1005-H-01L-VAC/0#
COMPARE
±3-2-3% full-scale (ANSI/ASME Grade B) 2 30 to 0\" Hg Utility Gauge, 2\" Dial, Bottom Connection
£13.82GBP / Each
£16.58GBP (incl VAT)
Ashcroft Utility Gauge, 0 to 30 psi, 2" Dial, Bottom Connection WZ-68003-02 Mfr # 20-W-1005-H-01L-30#
COMPARE
±3-2-3% full-scale (ANSI/ASME Grade B) 2 0 to 30 psi Utility Gauge, 2\" Dial, Bottom Connection
£12.82GBP / Each
£15.38GBP (incl VAT)
Ashcroft Utility Gauge, 0 to 60 psi, 2" Dial, Bottom Connection WZ-68003-03 Mfr # 20-W-1005-H-01L-60#
COMPARE
±3-2-3% full-scale (ANSI/ASME Grade B) 2 0 to 60 psi Utility Gauge, 2\" Dial, Bottom Connection
£37.07GBP / Each
£44.48GBP (incl VAT)
Ashcroft Utility Gauge, 0 to 100 psi, 2" Dial, Bottom Connection WZ-68003-04 Mfr # 20-W-1005-H-01L-100#
COMPARE
±3-2-3% full-scale (ANSI/ASME Grade B) 2 0 to 100 psi Utility Gauge, 2\" Dial, Bottom Connection
£12.82GBP / Each
£15.38GBP (incl VAT)
Ashcroft Utility Gauge, 0 to 160 psi, 2" Dial, Bottom Connection WZ-68003-05 Mfr # 20-W-1005-H-01L-160#
COMPARE
±3-2-3% full-scale (ANSI/ASME Grade B) 2 0 to 160 psi Utility Gauge, 2\" Dial, Bottom Connection
£12.82GBP / Each
£15.38GBP (incl VAT)
Ashcroft Utility Gauge, 0 to 200 psi, 2" Dial, Bottom Connection WZ-68003-06 Mfr # 20-W-1005-H-01L-200#
COMPARE
±3-2-3% full-scale (ANSI/ASME Grade B) 2 0 to 200 psi Utility Gauge, 2\" Dial, Bottom Connection
£12.82GBP / Each
£15.38GBP (incl VAT)
Ashcroft Utility Gauge, 30 to 0" Hg, 2 1/2" Dial, Bottom Connection WZ-68004-00 Mfr # 25-W-1005-H-02L-30IHGV
COMPARE
±3-2-3% full-scale (ANSI/ASME Grade B) 2.5 30 to 0\" Hg Utility Gauge, 2 1/2\" Dial, Bottom Connection
£14.73GBP / Each
£17.68GBP (incl VAT)
Ashcroft Utility Gauge, 0 to 15 psi, 2 1/2" Dial, Bottom Connection WZ-68004-01 Mfr # 25-W-1005-H-02L-15#
COMPARE
±3-2-3% full-scale (ANSI/ASME Grade B) 2.5 0 to 15 psi Utility Gauge, 2 1/2\" Dial, Bottom Connection
£14.73GBP / Each
£17.68GBP (incl VAT)
Ashcroft Utility Gauge, 0 to 30 psi, 2 1/2" Dial, Bottom Connection WZ-68004-02 Mfr # 25-W-1005-H-02L-30#
COMPARE
±3-2-3% full-scale (ANSI/ASME Grade B) 2.5 0 to 30 psi Utility Gauge, 2 1/2\" Dial, Bottom Connection
£13.82GBP / Each
£16.58GBP (incl VAT)
Ashcroft Utility Gauge, 0 to 100 psi, 2 1/2" Dial, Bottom Connection WZ-68004-04 Mfr # 25-W-1005-H-02L-100#
COMPARE
±3-2-3% full-scale (ANSI/ASME Grade B) 2.5 0 to 100 psi Utility Gauge, 2 1/2\" Dial, Bottom Connection
£13.82GBP / Each
£16.58GBP (incl VAT)
Ashcroft Utility Gauge, 0 to 160 psi, 2 1/2" Dial, Bottom Connection WZ-68004-05 Mfr # 25-W-1005-H-02L-160#
COMPARE
±3-2-3% full-scale (ANSI/ASME Grade B) 2.5 0 to 160 psi Utility Gauge, 2 1/2\" Dial, Bottom Connection
£13.82GBP / Each
£16.58GBP (incl VAT)
Ashcroft Utility Gauge, 0 to 200 psi, 2 1/2" Dial, Bottom Connection WZ-68004-06 Mfr # 25-W-1005-H-02L-200#
COMPARE
±3-2-3% full-scale (ANSI/ASME Grade B) 2.5 0 to 200 psi Utility Gauge, 2 1/2\" Dial, Bottom Connection
£13.82GBP / Each
£16.58GBP (incl VAT)
Ashcroft Utility Gauge, 0 to 300 psi, 2 1/2" Dial, Bottom Connection WZ-68004-07 Mfr # 25-W-1005-H-02L-300#
COMPARE
±3-2-3% full-scale (ANSI/ASME Grade B) 2.5 0 to 300 psi Utility Gauge, 2 1/2\" Dial, Bottom Connection
£13.82GBP / Each
£16.58GBP (incl VAT)
Ashcroft Utility Gauge, 0 to 400 psi, 2 1/2" Dial, Bottom Connection WZ-68004-08 Mfr # 25-W-1005-H-02L-400#
COMPARE
±3-2-3% full-scale (ANSI/ASME Grade B) 2.5 0 to 400 psi Utility Gauge, 2 1/2\" Dial, Bottom Connection
£13.82GBP / Each
£16.58GBP (incl VAT)
Showing 1- 20 of 20